missing translation for 'onlineSavingsMsg'
Learn More

Universal Indicator, Bogens in Alcohol, Certified, LabChem™

Supplier:  LabChem LC265007

 View more versions of this product

Catalog No. LC265007

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Universal Indicator
70,29.74,0.08,0.07,0.05
C27H28Br2O5S
Soluble in water
624.38
White Poly
Green
Liquid
Bogens in Alcohol
7732-18-5 , 76-59-5 , 76-16-9 , 547-58-0
UN1993
Passes Test
Certified
Carbon dioxide; Carbon monoxide; Sulfur compounds; Hydrogen bromide
125 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions