missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ UHPLC-MS Reagents Installation Kit
GREENER_CHOICE

Catalog No. UHPLCMSKIT
Change view
Click to view available options
Quantity:
6 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
UHPLCMSKIT 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. UHPLCMSKIT Supplier Thermo Scientific™ Supplier No. UHPLCMSKIT
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

  • The installation kit includes the following reagents:
    • Two (2)  bottles of W8-1, Water, UHPLC-MS grade
    • Two (2) bottles of A458-1, Methanol, UHPLC-MS grade
    • One (1) bottle of A956-1, Acetonitrile, UHPLC-MS grade
    • One (1) bottle of T111101000, ChromaCare™ LC-MS Instrument Flush Solution
  • All reagents in the kit are packaged in 1L borosilicate glass bottles
  • By reducing background noise, the T111101000 ChromaCare™ flush solution facilitates instrument installation and preventative maintenance routines
  • All reagents in the kit are available for individual sale

Specifications

Quantity 6 x 1 L
Green Features ACT Ecolabel (Silver)

RUO – Research Use Only