missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ IsoClean™ Front Snap Cleanroom Frocks, High Mandarin Collar
GREENER_CHOICE

Catalog No. 17988076
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17988076 X-Large
17988073 Small
17988074 Medium
17988075 Large
17988077 2X-Large
17988078 3X-Large
17988079 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.
Catalog No. 17988076 Supplier DuPont™ Supplier No. IC263SWHXL00300B
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Garments made of Tyvek™ should not be used around heat, flames, sparks or in potentially flammable or explosive environments.

Cleanroom frocks feature a high mandarin collar with snap for a secure fit and provides an inherent barrier to particles, microorganisms, and non-hazardous light liquid splash.

  • Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam
  • Elastic opening for tighter fit at wrist
  • Mandarin collar with snap for secure fit
  • Frocks provide body-length jacket design with front snap closures for easy donning and doffing
  • Bulk packaged in double transparent poly liners.
  • Manufactured under specifications that do not contain natural rubber latex

Specifications

Size X-Large
Material Tyvek
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type High Mandarin
Size (English) Chest 38 to 40 in.
Sleeve Style Set-in Sleeve
Length (Metric) 102.87 cm
Length (English) Sleeve 38 in.
Length (Metric) Sleeve 96.52 cm
Cleanroom Class Class 1000 (ISO 6)
Length (English) 40.5 in.
Product Type Controlled Environments Frock
Seam Type Serged
For Use With (Application) Cleanroom, Pharmaceutical, Biotechnology, Medical Device Manufacturing, and Laboratory Environments
Show More Show Less