Learn More
DuPont™ Tyvek™ 400 Lab Coat
GREENER_CHOICE
Catalog No. 17500278
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17500278 X-Large
17500264 2X-Large
17500266 3X-Large
17500268 4X-Large
17500273 Large
17500275 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17500278 Supplier DuPont™ Supplier No. TY212SWHXL0008G1
Limited-use Tyvek 400 lab coats provide protection and durability. Meets USDA, FDA, OSHA Standards.
  • Labcoat offers front snap closures for easy donning and doffing
  • Provides enhanced frontal protection
  • Laydown collar
  • Open wrists
  • Two front pockets for holding writing instruments, small tools or parts
  • Manufactured under specifications that do not contain natural rubber latex
Compliance
USDA, FDA, OSHA
Specifications
Product Type General Purpose Lab Coat
Size X-Large
Gender Unisex
Material Tyvek
Color White
Closure Type Snaps
Cuff Style Open
Sterility Non-sterile
Collar Type Plain
Seam Type Serged
Certifications/Compliance USDA, FDA, OSHA
Show More Show Less