missing translation for 'onlineSavingsMsg'
Learn More

Tyrphostin A1 98.0+%, TCI America™

Numéro de catalogue. T30741G
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
T30741G 1 g
T3074200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. T30741G Fournisseur TCI America Code fournisseur T30741G
Il en reste null

Tyrphostin A1 98.0+%, TCI America™ available now.

Spécifications

CAS 2826-26-8
Molecular Formula C11H8N2O
Synonym Tyrphostin 1, (4-Methoxybenzylidene)malononitrile
Quantity 1 g