missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 Coverall (all)

Catalog No. 19715336
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Seam Type:
Bound
Taped
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Seam Type
19715336 2X-Large Taped
19162778 5X-Large Taped
19715335 Large Taped
189992583 4X-Large Bound
19715334 Medium Taped
18760D X-Large Bound
18760E 3X-Large Bound
19162777 4X-Large Taped
18760C Large Bound
19087309 2X-Large Bound
18760B Medium Bound
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 19715336 Supplier DuPont™ Supplier No. SL125TWH2X000600

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful when handling many wet and dry chemicals, and certain hazardous materials such as PCBs - See more at: http://beta.fishersci.com/shop/products//p-25013#sthash.pbR5bBoS.dpuf

  • Pin locking slider locks in place when the pins on the puller are pushed into the zipper elements
  • Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
  • Laydown collar
  • Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
  • Elastic opening for tighter fit at wrist
  • Elastic opening for tighter fit at ankle
  • Manufactured under specifications that do not contain natural rubber latex

Compliance

Meets ASTM™ D3776, ASTM D1777, ASTM D3786, ASTM D5034 standards.

Specifications

Product Type Chemical Protective Clothing
Size 2X-Large
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Standard Collar
Closure Type Front Zipper
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals, Non-toxic Particulates
Coating Saranex 23-P film
Size (English) Chest 44.25 to 47.75 in.
Size (English) Inseam 30 in.
Size (Metric) Chest 112.39 to 121.28 cm
Size (Metric) Inseam 76.2 cm
For Use With (Application) Chemical mixing, hazardous remediation, emergency medical response, paint spraying and nuclear environments
Length (English) Sleeve 38.25 in.
Length (Metric) Sleeve 97.15 cm
Width (English) Chest 28.75 in.
Height (English) 6 to 6.4 ft.
Height (Metric) 1.82 to 1.95 m
Show More Show Less