missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Silvershield
Description
- 10000 Encapsulated Level A Suit with silvershield certified to NFPA 1990 (NFPA 1994, Class 2) 2022 edition
- Attached internal gloves composed of five layer laminate film for a broad range of chemical protection
- Neoprene gloves provide mechanical and physical durability to help protect inner gloves
- Integrated socks composed of garment material
- Attached flaps are designed to cover boot tops to help reduce potential for liquid intrusion
- Two exhaust valves release higher pressure inside encapsulating garments while simultaneously helping to prevent inward leakage of external vapors or particles into the garment
- One-way valves are positive pressure and open on demand
- Include splash covers made of barrier material to help prevent liquid intrusion
- Located on left back of head and right lower back
- Internal adjustable waist belt system for support and improved fit
- Each suit has a unique serial number and is fully tested at time of manufacture including positive air pressure integrity testing per ASTM F1052
- Manufactured under specifications that do not contain natural rubber latex
- Made in the USA, North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
- Class 2 garments are designed for situations where there is an immediate danger to life and health (IDLH) and must be worn with a self-contained breathing apparatus (SCBA)
- Encapsulated Level A garment design is our highest level of protection from liquid splash and vapor/gas exposures for both the wearer and respiratory equipment
- Double taped seams provide strong chemical resistance against heavy liquid splashes
- Sewn seam is covered, both inside and outside of garment with compatible chemical-resistant material by heat-sealing.
- Expanded anti-fog face shield provides wide 220° viewing angle
- EX face shield is three-layer laminate composed of 40 mil PVC, 5 mil Teflon™, 20 mil PVC
- Front entry design allows wearer to partially don garment (maintain standby position) without assistance
- Expanded back to accommodate self-contained air breathing apparatus (SCBA)
- Vapor tight zipper
- Extra long to increase garment opening and aid donning and doffing
- Storm flap covers zipper which can be secured by the wearer with rugged hook and loop material to prevent intrusion at zipper
Certifications
NFPA 1994 Class 2
Specifications
Specifications
| Color | Lime Yellow |
| For Use With (Application) | Industrial, Chemical Handling, Emergency Response |
| Material | Tychem |
| Size | Medium |
| Ankle Style | Boot Overflaps |
| Closure Type | Zipper |
| Product Type | Encapsulated Level A Suit |
| Seam Type | Taped |