missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Turbidity Standard, 10 NTU, Polymer Bead Suspension, Ricca Chemical
Supplier: Ricca Chemical Company R88010004C
Specifications
| Turbidity Standard | |
| Amber Glass Bottle | |
| 98.0,0.02,0.01 | |
| Liquid | |
| MFCD00084450 | |
| 100°C | |
| *-CC(-*)C1=CC=CC=C1 | |
| 104.15 | |
| 7501 |
| 10 NTU (Nephelometric Turbidity Units) | |
| 9003-53-6 , 26628-22-8 , 7732-18-5 | |
| 100.0,0.02,0.01 | |
| (C8H8)n | |
| Colorless | |
| PPBRXRYQALVLMV-UHFFFAOYSA-N | |
| ethenylbenzene | |
| 4 L | |
| CHEBI:27452 |
Chemical Identifiers
| 9003-53-6 | |
| 104.15 | |
| PPBRXRYQALVLMV-UHFFFAOYSA-N | |
| CHEBI:27452 | |
| *-CC(-*)C1=CC=CC=C1 |
| (C8H8)n | |
| MFCD00084450 | |
| 7501 | |
| ethenylbenzene |