CAS RN 26628-22-8
Sodium azide, 99%, extra pure
CAS: 26628-22-8 MDL Number: MFCD00003536 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Sodium Azide, 5% (w/v), Ricca Chemical
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.011 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Sodium Azide, Crystalline, Laboratory, Fisher Chemical™
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.011 MDL Number: MFCD00003536 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Sodium Azide, White Powder, Fisher BioReagents
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.011 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Thermo Scientific Chemicals Sodium azide, 99+%, for biochemistry
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.01 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Protein Standard Mix 15 - 600 kDa for size exclusion chromatography, MilliporeSigma™ Supelco™
The protein standard mix is a calibration standard to test and monitor performance of size exclusion chromatography (SEC) columns. It is a lyophilized mixture of molecular weight markers ranging from 15 kDa to 600 kDa.
Sodium Azide, ≥99%
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.011 MDL Number: MFCD00003536 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Sodium azide, 99%
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.011 MDL Number: MFCD00003536 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Chemical Solution, Buffer Solution pH 10.0 at 25°C, Color Coded Blue, Traceable to NIST, LabChem
This Chemical Solution, Buffer Solution has a pH of 10.0 at 25°C, is Color Coded Blue, and is Traceable to NIST.
Sodium Azide, For Chromium, 0.5% (w/v), LabChem™
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.011 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Sodium Azide 99.0+%, TCI America™
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.011 MDL Number: MFCD00003536 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N Synonym: sodium azide,azide, sodium,natriumazid,nemazyd,smite,kazoe,azoture de sodium,sodium azide na n3,hydrazoic acid, sodium salt,natriumazid german PubChem CID: 33557 ChEBI: CHEBI:278547 IUPAC Name: sodium;azide SMILES: [N-]=[N+]=[N-].[Na+]
Sodium Azide, Granular, 99%, Spectrum™ Chemical
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.01 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N IUPAC Name: sodium azide SMILES: [Na+].[N-]=[N+]=[N-]
Sodium Azide, ACS, 99%, Spectrum™ Chemical
CAS: 26628-22-8 Molecular Formula: N3Na Molecular Weight (g/mol): 65.01 InChI Key: PXIPVTKHYLBLMZ-UHFFFAOYSA-N IUPAC Name: sodium azide SMILES: [Na+].[N-]=[N+]=[N-]