missing translation for 'onlineSavingsMsg'
Learn More

Turbidity Standard, 10 NTU, Polymer Bead Suspension, Ricca Chemical

Catalog No. p-3252601
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
Clear PET-G Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R8801000500 500 mL Clear PET-G Bottle
R88010004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R8801000500 Supplier Ricca Chemical Company Supplier No. R8801000500G
Only null left

Chemical Identifiers

CAS 9003-53-6 , 26628-22-8 , 7732-18-5
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name ethenylbenzene
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Packaging Clear PET-G Bottle
CAS Min % 98.0, 0.02, 0.01
CAS Max % 100.0, 0.02, 0.01
Physical Form Liquid
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Chemical Name or Material Turbidity Standard, 10 NTU (Nephelometric Turbidity Units)