Learn More
United Scientific™ Tuning Pipes
These tuning pipes create a long-lasting sound that fills an entire room.
Supplier:  United Scientific™ TUNPIPSET3
 Afficher plus de versions de ce produit
Numéro de catalogue. S000864
Il en reste null
Avertissement:
Prop 65
Description
Description
Compared to traditional tuning forks, they are much louder and have a more penetrating tone. Made from an aluminum alloy, each pipe is tuned to a specific frequency. The tuning pipe is supported by a wooden handle and string, allowing it to vibrate freely without any dampening from being held. The frequency is laser etched onto each pipe for easy reference. For best results, a rubber hammer is included to activate the pipe.
  • Carefully tuned to each frequency
  • Made from high quality, heat-treated aluminum
  • Louder than traditional tuning forks, with a long-lasting sound
  • Available in 256 Hz., 384 Hz., and 528 Hz., and as a set of three
Spécifications
Identifiants chimiques
Tuning Pipe
Spécifications
Tuning Pipe
256 Hz, 384 Hz, 582 Hz
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS