Learn More
United Scientific™ Tuning Pipes
These tuning pipes create a long-lasting sound that fills an entire room.
$190.21 - $539.75
| Catalog Number | Mfr. No. | Frequency | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Frequency | Price | Quantity & Availability | |||||
|
S000880
|
United Scientific™
TUNPIP256 |
256 Hz |
Each for $190.21
|
|
|||||
|
S000881
|
United Scientific™
TUNPIP384 |
384 Hz |
Each for $190.21
|
|
|||||
|
S000863
|
United Scientific™
TUNPIP528 |
582 Hz |
Each for $190.21
|
|
|||||
|
S000864
|
United Scientific™
TUNPIPSET3 |
256 Hz, 384 Hz, 582 Hz |
Each for $539.75
|
|
|||||
Description
Compared to traditional tuning forks, they are much louder and have a more penetrating tone. Made from an aluminum alloy, each pipe is tuned to a specific frequency. The tuning pipe is supported by a wooden handle and string, allowing it to vibrate freely without any dampening from being held. The frequency is laser etched onto each pipe for easy reference. For best results, a rubber hammer is included to activate the pipe.
- Carefully tuned to each frequency
- Made from high quality, heat-treated aluminum
- Louder than traditional tuning forks, with a long-lasting sound
- Available in 256 Hz., 384 Hz., and 528 Hz., and as a set of three
Specifications
| Tuning Pipe |