missing translation for 'onlineSavingsMsg'
Learn More

Tungsten Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC PLW92M

 View more versions of this product

Catalog No. PLW92M


Only null left
Explore available promotions

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
W1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Tungsten Standard
7440-33-7 , 7732-18-5
0.1,99.9
W
AA, ICP
Certified Reference Material
[W]
tungsten
30 mL
CHEBI:27998
Matrix    Concentration    Cat. No.
H2O    1000μg/mL    PLW9-2Y, -2X    10,000    PLW9-3Y, -3X
1% HNO3
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,99.9
MFCD00011461
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
WFKWXMTUELFFGS-UHFFFAOYSA-N
Water
183.84
23964
Assurance

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
CHEBI:27998
[W]
W
MFCD00011461
23964
tungsten
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only