missing translation for 'onlineSavingsMsg'
Learn More

Tungsten Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$143.43 - $278.50

Chemical Identifiers

CAS 7440-33-7
Molecular Formula W
Molecular Weight (g/mol) 183.84
MDL Number MFCD00011461
InChI Key WFKWXMTUELFFGS-UHFFFAOYSA-N
PubChem CID 23964
ChEBI CHEBI:27998
IUPAC Name tungsten
SMILES [W]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLW92M
View Documents
SPEX CertiPrep Group LLC
PLW92M
30 mL Triple-leached LDPE bottle
Each for $143.43
Only null left
 
PLW9-2Y
View Documents
SPEX CertiPrep Group LLC
PLW92Y
125 mL Triple-leached LDPE bottle
Each for $278.50
Only null left
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
W1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7440-33-7
183.84
WFKWXMTUELFFGS-UHFFFAOYSA-N
CHEBI:27998
[W]
W
MFCD00011461
23964
tungsten

Specifications

Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
0.1,99.9
MFCD00011461
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
WFKWXMTUELFFGS-UHFFFAOYSA-N
Water
183.84
23964
Assurance
Tungsten Standard
7440-33-7 , 7732-18-5
0.1,99.9
W
AA, ICP
Certified Reference Material
[W]
tungsten
30 mL
CHEBI:27998
Matrix    Concentration    Cat. No.
H2O    1000μg/mL    PLW9-2Y, -2X    10,000    PLW9-3Y, -3X
1% HNO3
Videos
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient

SDS
Documents

Documents

Product intended for laboratory use only