CAS RN 7440-33-7
Tungsten slug, 3.175mm (0.125 in.) dia. x 6.35mm (0.25 in.) length, 99.97% (metals basis), Thermo Scientific Chemicals
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten powder, APS <1 micron, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten sputtering target, 76.2mm (3.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.95% (metals basis)
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten gauze, 150 mesh woven from 0.02mm (0.0008in) dia wire
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten wire, 0.015mm (0.00059in) dia, 99.95% (metals basis)
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten wire basket; # turns, 8; ID(mm), 7; Ht(mm), 9; Wire leads(cm), 5.0; Wire dia(mm), 0.51, Thermo Scientific™
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten gauze, 20 mesh woven from 0.127mm (0.005in) dia wire
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten gauze, gold plated, 50 mesh woven from 0.025mm (0.001in) dia wire
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten wire basket; # turns, 9; ID(mm), 4; Ht(mm), 7; Wire leads(cm), 3.5; Wire dia(mm) 0.51, Thermo Scientific™
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten gauze, 100 mesh woven from 0.0509mm (0.002in) dia wire
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten gauze, 30 mesh woven from 0.1018mm (0.004in) dia wire
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]
Tungsten sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
CAS: 7440-33-7 Molecular Formula: W Molecular Weight (g/mol): 183.84 MDL Number: MFCD00011461 InChI Key: WFKWXMTUELFFGS-UHFFFAOYSA-N Synonym: wolfram,ion,va,tungsten, elemental,wolframium,unii-v9306cxo6g,4+ ion,tungsten, soluble compounds,tungsten, insoluble compounds,powder PubChem CID: 23964 ChEBI: CHEBI:27998 IUPAC Name: tungsten SMILES: [W]