missing translation for 'onlineSavingsMsg'
Learn More

Wanzl TSS Clear Basket Label Holder

Numéro de catalogue. 11222123
Change view
Click to view available options
Width (English):
24 in.
36 in.
48 in.
60 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Width (English)
11222123 60 in.
11222120 24 in.
11222121 36 in.
11222122 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. 11222123 Fournisseur Wanzl Code fournisseur T.TLB60CL
Il en reste null

Clear Basket Label Holder for use with TSS Shelving System.

  • Add-on option for additional customization
  • Clear
  • Three ways to mount

Spécifications

For Use With (Application) Shelving
Width (English) 60 in.
Color Clear
Includes Basket Label Holder
Material Plastic