missing translation for 'onlineSavingsMsg'
Learn More

Wanzl TSS Chrome 4 Basket Rolling Unit with Solid Base

Numéro de catalogue. 11222098
Change view
Click to view available options
Dimensions (L x W):
18 x 36 in.
18 x 48 in.
18 x 60 in.
24 x 36 in.
24 x 48 in.
24 x 60 in.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Dimensions (L x W)
11222098 24 x 48 in.
11222094 18 x 36 in.
11222095 18 x 48 in.
11222096 18 x 60 in.
11222097 24 x 36 in.
11222099 24 x 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. 11222098 Fournisseur Wanzl Code fournisseur T.MBSK2448CHSLD
Il en reste null

TSS by Wanzl is recognized for high quality and strength due to our all-welded construction and truss configuration. This system requires no tools for assembly, making the process simple and easy to follow.

  • Adjustable at 1 in. increments and can be utilized with shelves and other accessories to create a custom storage unit.
  • Shallow hopper front allows for easy access to items.
  • Dividers can be added to compartmentalize
  • Offered in our Chrome Plus (CH) finish. The heavy nickel plate process features a hard chromium finish with special brighteners to enhance and increase the life of the shelving.

Spécifications

Includes 4 Basket Unit
Width (English) 48 in.
Dimensions (L x W) 24 x 48 in.
For Use With (Application) Shelving
Color Chrome
Material Steel