missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TrueCut™ HiFi Cas9 Protein (1 μg/μL)

Catalog No. p-7235828
Encompass
Change view
Click to view available options
Quantity:
10 μg
25 μg
Concentration:
1 μg/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
A50574 10 μg 1 μg/μL
A50575 25 μg 1 μg/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50574 Supplier Invitrogen™ Supplier No. A50574

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A high fidelity CRISPR Cas9 protein variant engineered to demonstrate superior off-target profiles, while preserving maximum on-target editing efficiency. TrueCut™ HiFi Cas9 Protein is ideal for applications that are sensitive to off-target events and when more precise editing is required.

  • A high fidelity CRISPR Cas9 protein variant engineered to demonstrate superior off-target profiles, while preserving maximum on-target editing efficiency
  • Near complete elimination of off-target events in all tested cell lines including standard, immune, primary, and stem
  • Maintains maximum on-target editing efficiency compared to TrueCut™ Cas9 Protein v2
  • Transfection-ready using either electroporation or lipid-mediated transfection reagents
  • Manufactured under strict ISO-13485 quality standards
  • Ideal for applications that are sensitive to off-target events and when more precise editing is required
  • Eliminates time consuming cloning steps

Available in Two Concentrations

  • 1 μg/μL - For use in standard editing scenarios
  • 5 μg/μL - For optimized editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time
  • For custom sizes or concentrations, please inquire

Specifications

Concentration 1 μg/μL
Purity ≥90%
Sensitivity Yes
Source E. coli
Quantity 10 μg
Product Type HiFi Cas9
Form Liquid
How is Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) different from Invitrogen TrueCut HiFi Cas9 Protein (Cat. No. A50574, A50575)?

Both Gibco CTS HiFi Cas9 Protein (Cat. No. A54223, A54224) and Invitrogen TrueCut HiFi Cas9 Protein (Cat. No. A50574, A50575) are Streptococcus pyogenes high-fidelity CRISPR Cas9 proteins, produced by recombinant DNA technology in E. coli. Both proteins have the same sequence with added nuclear localization signal. Gibco CTS HiFi Cas9 Protein is manufactured to meet the standards for “Ancillary Materials for Cell, Gene, and Tissue-Based Products” whereas Invitrogen TrueCut HiFI Cas9 Protein is a research-grade product.

Do you offer TrueCut HiFi Cas9 Protein at different concentrations?

Yes, we offer TrueCut HiFi Cas9 Protein in the following concentrations:
1 µg/µL (Cat. Nos. A50574, A50575) for use in standard editing scenarios 5 µg/µL (Cat. Nos. A50576, A50577) for optimization of editing conditions in more difficult scenarios such as in primary or embryonic cells, microinjection, or when screening multiple gRNA sequences at a time

What is the difference between TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?

TrueCut HiFi Cas9 Protein is a modified Cas9 enzyme engineered to exhibit superior off-target profile, while maintaining maximum on-target cleavage efficiency of the wild type TrueCut Cas9 Protein v2. TrueCut HiFi Cas9 Protein is ideal for experiments that are sensitive to off-target events while still requiring a high level of editing efficiency.

What is the storage buffer composition for TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?

The storage buffer composition for TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein is as follows:
15 mM Tris-HCl pH 8.0, 150 mM NaCl, 1.0 mM TCEP, 50% glycerol.

Which lipid transfection reagent do you recommend using with TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?

We recommend using Lipofectamine CRISPRMAX Transfection Reagent with TrueCut Cas9 Protein v2 or TrueCut HiFi Cas9 Protein.

What gRNA format do you recommend using with TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein?

TrueCut Cas9 Protein v2 and TrueCut HiFi Cas9 Protein are designed to work with TrueGuide synthetic gRNAs, but can also be used with in vitro transcribed gRNA from the GeneArt Precision gRNA Synthesis Kit.