missing translation for 'onlineSavingsMsg'
Learn More

Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.), Fisher BioReagents™

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

Supplier:  Fisher BioReagents BP1758100

 View more versions of this product

Catalog No. BP1758-100



Only null left

Description

Description

Tris Hydrochloride, 1M Solution (pH 8.0/Mol. Biol.)

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 8.0 provide convenient and highly purified solutions.
Specifications

Specifications

Tris Hydrochloride
77-86-1 , 1185-53-1 , 7732-18-5
0.96 to 1.04 M
Liquid
DNase free
DNase-, RNase- and Protease-Free
C4H11NO3
Protease free
LENZDBCJOHFCAS-UHFFFAOYSA-N
2-amino-2-(hydroxymethyl)propane-1,3-diol
6503
Molecular Biology
1M Solution, pH 8.0
Undesignated
10
100 mL
Filtered through a 0.2-micron filter.
Pass Test
Poly Bottle
Tris-HCl
C(C(CO)(CO)N)O
121.136
CHEBI:9754

Chemical Identifiers

77-86-1
121.136
Tris-HCl
CHEBI:9754
C(C(CO)(CO)N)O
C4H11NO3
LENZDBCJOHFCAS-UHFFFAOYSA-N
6503
2-amino-2-(hydroxymethyl)propane-1,3-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions