missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris Hydrochloride, 1M Solution (pH 7.5/Mol. Biol.), Fisher BioReagents™
Supplier: Fisher BioReagents BP1757500
Description
Tris Hydrochloride, 1M Solution (pH 7.5/Mol. Biol.)
Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 7.5 provide convenient and highly purified solutions.Specifications
| Tris Hydrochloride | |
| 77-86-1 , 1185-53-1 , 7732-18-5 | |
| 0.96 to 1.04 M | |
| Liquid | |
| DNase free | |
| DNase-, RNase- and Protease-Free | |
| C4H11NO3 | |
| Protease free | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol | |
| 6503 | |
| Molecular Biology |
| 1M Solution, pH 7.5 | |
| Undesignated | |
| 7.5 | |
| 500 mL | |
| Filtered through a 0.2-micron filter. | |
| Pass Test | |
| Poly Bottle | |
| Tris-HCl | |
| C(C(CO)(CO)N)O | |
| 121.136 | |
| CHEBI:9754 |
Chemical Identifiers
| 77-86-1 | |
| 121.136 | |
| Tris-HCl | |
| CHEBI:9754 | |
| C(C(CO)(CO)N)O |
| C4H11NO3 | |
| LENZDBCJOHFCAS-UHFFFAOYSA-N | |
| 6503 | |
| 2-amino-2-(hydroxymethyl)propane-1,3-diol |
Safety and Handling
Recommended Storage : RT