missing translation for 'onlineSavingsMsg'
Learn More

Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.), Fisher BioReagents™

Catalog No. p-29023
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1756100 100 mL Poly Bottle
BP1756500 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1756100 Supplier Fisher BioReagents Supplier No. BP1756 100
Only null left

Tris Hydrochloride, 1M Solution (pH 7.0/Mol. Biol.)

Tris is a buffer component in molecular biology, tissue culture, and electrophoresis procedures. These 1M solutions of Tris Hydrochloride at pH 7.0 provide convenient and highly purified solutions.

Chemical Identifiers

CAS 77-86-1 , 1185-53-1 , 7732-18-5
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Tris-HCl
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Color Undesignated
Concentration 0.96 to 1.04 M
pH 10
Physical Form Liquid
Quantity 100 mL
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Identification Pass Test
Packaging Poly Bottle
Protease Protease free
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Hydrochloride, 1M Solution, pH 7.0
Show More Show Less
Recommended Storage RT