missing translation for 'onlineSavingsMsg'
Learn More
Learn More
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
18-048-033
|
Medisca Pharmaceutique Inc
077004 |
25 g |
Each for $95.07
|
|
|||||
|
18-048-034
|
Medisca Pharmaceutique Inc
077009 |
1 kg |
Each for $1,502.75
|
|
|||||
|
18-048-092
|
Medisca Pharmaceutique Inc
077000* |
10 kg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18-048-137
|
Medisca Pharmaceutique Inc
077005 |
100 g |
Each for $239.22
|
|
|||||
Description
Trimethoprim USP can be white to cream-colored, odorless crystals or crystalline powder. Particles micronized so that 90% are less than 20μm.
Chemical Identifiers
| 738-70-5 |
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| 738-70-5 | |
| 25 g | |
| USP |
| Crystalline Powder | |
| Cream to White | |
| 98.5 to 101.0% |