missing translation for 'onlineSavingsMsg'
Learn More

Trimethoprim USP, Medisca™
GREENER_CHOICE

Catalog No. 18048033
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18048033 25 g
18048034 1 kg
18048092 10 kg
18048137 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18048033 Supplier Medisca Pharmaceutique Inc Supplier No. 077004

Trimethoprim USP can be white to cream-colored, odorless crystals or crystalline powder. Particles micronized so that 90% are less than 20μm.

Chemical Identifiers

CAS 738-70-5

Specifications

Type α-Carboxybenzyl Penicillin Disodium Salt
Physical Form Crystalline Powder
Color Cream to White
Quantity 25 g
Percent Purity 98.5 to 101.0%
Grade USP