accessibility menu, dialog, popup

UserName

H3CONNH2NH2NOH3COH3C

CAS RN 738-70-5

IUPAC Name: 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Synonyms: trimethoprim abacin alprim sulfatrim bactrim DS imexim sigaprim bactin ipral eusaprim
Molecular Weight (g/mol): 290.32
Molecular Formula: C14H18N4O3
InChi Key: IEDVJHCEMCRBQM-UHFFFAOYSA-N
SMILES: COC1=CC(CC2=CN=C(N)N=C2N)=CC(OC)=C1OC
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (4)
Physical Form 
  • (1)
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Type 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (4)
  • (1)
  • (1)
  • (8)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (2)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (4)
  • (4)
  • (3)