Learn More
Trimethoprim USP, Medisca™
Supplier: Medisca Pharmaceutique Inc 077000*
Description
Trimethoprim USP can be white to cream-colored, odorless crystals or crystalline powder. Particles micronized so that 90% are less than 20μm.
Specifications
| α-Carboxybenzyl Penicillin Disodium Salt | |
| Cream to White | |
| 98.5 to 101.0% |
| 738-70-5 | |
| 10 kg | |
| USP |
Chemical Identifiers
| 738-70-5 |