missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethoxysilane, 96%
Description
Triethoxysilane is raw material for synthesis of silane addition agent and silicon oil. It is used as a reducing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethoxysilane is raw material for synthesis of silane addition agent and silicon oil. It is used as a reducing agent.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Moisture sensitive. Store away from strong oxidizing agents, strong acids, bases, water/ moisture. Store under inert gas.
Triethoxysilane is raw material for synthesis of silane addition agent and silicon oil. It is used as a reducing agent.
Solubility
Insoluble in water. Soluble in organic solvents.
Notes
Moisture sensitive. Store away from strong oxidizing agents, strong acids, bases, water/ moisture. Store under inert gas.
Spécifications
Spécifications
| Boiling Point | 134°C to 135°C |
| Melting Point | -170°C |
| Quantity | 10 g |
| Linear Formula | (CH3CH2O)3SiH |
| UN Number | UN3384 |
| Beilstein | 1738989 |
| Flash Point | 26°C (78°F) |
| Solubility Information | Insoluble in water. Soluble in organic solvents. |
| Formula Weight | 164.28 |
| Percent Purity | 96% |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | H226-H302-H315-H318-H330-H335 |
| Hazard Statement | GHS H Statement H330-H314-H318-H226 Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Flammable liquid and vapour. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P314-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S. |
| EINECSNumber | 213-650-7 |
| RTECSNumber | VV6682000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only