missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Translucent Narrow Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
The translucent narrow-mouth amber HDPE reagent bottles with polypropylene caps are UV-protective, leak-tested, and chemical-resistant, ideal for safe storage and transport of light-sensitive liquids in lab environments.
$43.07 - $161.50
Specifications
| Color | Amber |
|---|---|
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000092
|
Eisco Labs
CH5385PK12 |
4 mL |
Pack of 12 Each for $43.07
|
|
|||||
|
S000093
|
Eisco Labs
CH5386PK12 |
8 mL |
Pack of 12 Each for $46.68
|
|
|||||
|
S000094
|
Eisco Labs
CH5387PK12 |
15 mL |
Pack of 12 Each for $50.21
|
|
|||||
|
S000095
|
Eisco Labs
CH5388PK12 |
30 mL |
Pack of 12 Each for $57.39
|
|
|||||
|
S000096
|
Eisco Labs
CH5391PK12 |
250 mL |
Pack of 12 Each for $125.61
|
|
|||||
|
S000097
|
Eisco Labs
CH5392PK12 |
500 mL |
Pack of 12 Each for $161.50
|
|
|||||
Description
- Amber Reagent Bottles: Includes 12 sterile narrow-mouth HDPE bottles with polypropylene caps, ideal for secure sample storage and transportation.
- Light-Sensitive Sample Protection: Amber color provides shielding from UV light, perfect for storing sensitive reagents and solutions.
- Leak-Proof Tested Design: Tested under pressure to prevent leaks, ensuring safe storage and transport of liquid samples in labs and industrial settings.
- Durable And Chemical-Resistant: Constructed from HDPE, these bottles withstand a wide range of chemicals, making them suitable for diverse lab applications.
- Convenient Handling And Transport: Features a translucent body for easy visibility, handling, and portability in laboratory environments.
Specifications
| Amber | |
| 12 Pk. | |
| Yes | |
| Narrow | |
| Not Autoclavable |
| HDPE | |
| Sterile | |
| No | |
| Reagent Bottle |