missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Translucent Narrow Mouth Amber HDPE Reagent Bottles with Polypropylene Caps

The translucent narrow-mouth amber HDPE reagent bottles with polypropylene caps are UV-protective, leak-tested, and chemical-resistant, ideal for safe storage and transport of light-sensitive liquids in lab environments.

Supplier:  Eisco Labs CH5391PK12

 View more versions of this product

Catalog No. S000096


Only null left

Description

Description

  • Amber Reagent Bottles: Includes 12 sterile narrow-mouth HDPE bottles with polypropylene caps, ideal for secure sample storage and transportation.
  • Light-Sensitive Sample Protection: Amber color provides shielding from UV light, perfect for storing sensitive reagents and solutions.
  • Leak-Proof Tested Design: Tested under pressure to prevent leaks, ensuring safe storage and transport of liquid samples in labs and industrial settings.
  • Durable And Chemical-Resistant: Constructed from HDPE, these bottles withstand a wide range of chemicals, making them suitable for diverse lab applications.
  • Convenient Handling And Transport: Features a translucent body for easy visibility, handling, and portability in laboratory environments.
Specifications

Chemical Identifiers

Amber
12 Pk.
Yes
Narrow
Not Autoclavable
HDPE
Sterile
No
Reagent Bottle

Specifications

250 mL
Amber
HDPE
Sterile
No
133 mm
62 mm
8.45 oz.
17.6 mm
12 Pk.
Yes
Narrow
Reagent Bottle
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions