missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Translucent Narrow Mouth Amber HDPE Reagent Bottles with Polypropylene Caps

Catalog No. S000097
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
250 mL
500 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000097 500 mL
S000092 4 mL
S000093 8 mL
S000094 15 mL
S000095 30 mL
S000096 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S000097 Supplier Eisco Labs Supplier No. CH5392PK12
Only null left

The translucent narrow-mouth amber HDPE reagent bottles with polypropylene caps are UV-protective, leak-tested, and chemical-resistant, ideal for safe storage and transport of light-sensitive liquids in lab environments.

  • Amber Reagent Bottles: Includes 12 sterile narrow-mouth HDPE bottles with polypropylene caps, ideal for secure sample storage and transportation.
  • Light-Sensitive Sample Protection: Amber color provides shielding from UV light, perfect for storing sensitive reagents and solutions.
  • Leak-Proof Tested Design: Tested under pressure to prevent leaks, ensuring safe storage and transport of liquid samples in labs and industrial settings.
  • Durable And Chemical-Resistant: Constructed from HDPE, these bottles withstand a wide range of chemicals, making them suitable for diverse lab applications.
  • Convenient Handling And Transport: Features a translucent body for easy visibility, handling, and portability in laboratory environments.

Specifications

Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Amber
Diameter (Metric) Inner Neck 21.6 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 170 mm
Product Type Reagent Bottle
Diameter (Metric) 73.5 mm
Autoclavable Not Autoclavable
Show More Show Less