missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Translucent Narrow Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
The translucent narrow-mouth amber HDPE reagent bottles with polypropylene caps are UV-protective, leak-tested, and chemical-resistant, ideal for safe storage and transport of light-sensitive liquids in lab environments.
Supplier: Eisco Labs CH5388PK12
Description
- Amber Reagent Bottles: Includes 12 sterile narrow-mouth HDPE bottles with polypropylene caps, ideal for secure sample storage and transportation.
- Light-Sensitive Sample Protection: Amber color provides shielding from UV light, perfect for storing sensitive reagents and solutions.
- Leak-Proof Tested Design: Tested under pressure to prevent leaks, ensuring safe storage and transport of liquid samples in labs and industrial settings.
- Durable And Chemical-Resistant: Constructed from HDPE, these bottles withstand a wide range of chemicals, making them suitable for diverse lab applications.
- Convenient Handling And Transport: Features a translucent body for easy visibility, handling, and portability in laboratory environments.
Chemical Identifiers
| Amber | |
| 12 Pk. | |
| Yes | |
| Narrow | |
| Not Autoclavable |
| HDPE | |
| Sterile | |
| No | |
| Reagent Bottle |
Specifications
| 30 mL | |
| Amber | |
| HDPE | |
| Sterile | |
| No | |
| 62 mm | |
| 36.5 mm |
| 1 oz. | |
| 14 mm | |
| 12 Pk. | |
| Yes | |
| Narrow | |
| Reagent Bottle | |
| Not Autoclavable |