missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Indeno[1,2,3-cd]pyrene, MilliporeSigma™ Supelco™
Description
Environmental
Spécifications
Spécifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| Product Line | TraceCERT |
| CAS | 193-39-5 |
| Physical Form | Neat |
| Linear Formula | C22H12 |
| MDL Number | MFCD00152577 |
| Type | Certified Reference Material |
| Quantity | 10 mg |
| Formula Weight | 276.33 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC