missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ HPLC Gradient System Diagnostic Mix, Certified Reference Material, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Flash Point | 10°C (Closed Cup) |
| UN Number | UN1230 - Class 3 - PG 2 - Methanol, solution |
| Quantity | 6 x 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC