missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Azinphos-ethyl Certified Reference Material, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Product Line | TraceCERT |
| CAS | 2642-71-9 |
| Linear Formula | C12H16N3O3 PS2 |
| MDL Number | MFCD00055539 |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 345.38 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC