missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ 2,4-Dinitrotoluene Certified Reference Material, MilliporeSigma™ Supelco™
Spécifications
Spécifications
| Product Line | TraceCERT |
| CAS | 121-14-2 |
| Vapor Pressure | 1 mmHg (102.7°C) |
| Linear Formula | CH3C6H3(NO2)2 |
| MDL Number | MFCD00007172 |
| Melting Point | 67°C to 70°C |
| Type | Certified Reference Material |
| Quantity | 100 mg |
| Formula Weight | 182.13 |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC