missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ 1,4-Dioxane Solution, 2,000 μg/mL in Methanol, MilliporeSigma™ Supelco™

Numéro de catalogue. 111017908
Modifier l'affichage
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111017908 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111017908 Fournisseur Merck Emd Millipore Code fournisseur CRM48367
Il en reste null
Environmental

Spécifications

Packaging Ampule
Product Line TraceCERT
CAS 123-91-1
Linear Formula C4H8O2
MDL Number MFCD00006571
Synonym 1,4-Dioxane solution
Type Organic Pollutant Standard
Quantity 1 mL
Formula Weight 88.11
Grade TraceCERT™ Analytical Standard
Concentration or Composition (by Analyte or Components) 2,000 μg/mL in methanol
Afficher plus de résultats Afficher moins de résultats

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC