missing translation for 'onlineSavingsMsg'
Learn More

Ansell TouchNTuff™ 92-600 Disposable Chemical Resistant Nitrile Gloves

Catalog No. 189992748E
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
189992748E 2X-Large
189992748AA X-Small
189992748A Small
189992748B Medium
189992748C Large
189992748D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 189992748E Supplier Ansell Supplier No. 92600110
Only null left

Soft nitrile disposable gloves provide high levels of comfort and splash resistance against hazardous chemicals.

  • Made from 100% nitrile
  • Resistant to nuisance punctures, nicks, cuts and abrasions
  • Comfortable thin mil construction
  • Rolled cuffs for easy donning and doffing
  • Chemical resistant

Specifications

Applications Industrial
Color Green
Latex Free Yes
Material Nitrile
Product Type Chemical-resistant Gloves
Size 2X-Large
Glove Cuff Style Rolled
Quantity 900/Cs.