missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TNF alpha Uncoated ELISA Kit

ELISA

$1,082.00 - $1,476.00

Spécifications

Accession Number P01375
Assay Range 4 to 500 pg/mL
Assay Sensitivity 4 pg/mL
Conjugate HRP
Product Type ELISA
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
5017366
Afficher les documents Détails de la promotion
Invitrogen™
88734677
20 x 96 Tests
chaque for $1,476.00
Il en reste null
 
501128921
Afficher les documents Détails de la promotion
Invitrogen™
88734688
10 x 96 Tests
chaque for $1,082.00
Il en reste null
 
Description

Description

The Human Tumor Necrosis Factor Alpha (TNF-α) Uncoated ELISA Kit contains pre-matched antibody pairs,and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human TNF-α. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

TNF alpha is a multifunctional proinflammatory cytokine that belongs to the tumor necrosis factor (TNF) superfamily. This cytokine is mainly secreted by macrophage and bind to its receptors, TNFRSF1A/TNFR1 and TNFRSF1B/TNFBR. TNF alpha is involved in the regulation of immune cells, cell proliferation, differentiation, apoptosis, lipid metabolism, and coagulation. TNF alpha exists as a multimer of two, three, or five noncovalently linked units, but shows a single 17 kDa band following SDS-PAGE under non-reducing conditions. Knockout studies in mice also suggested the neuroprotective function of TNF alpha, and has been observed to causes tumor necrosis when injected into tumor-bearing mice. Other functions of TNF-alpha include its role in the immune response to bacterial, viral, parasitic and certain fungal infections, as well as its role in the necrosis of specific tumors. TNF alpha causes cytolysis or cytostasis of certain transformed cells, being synergistic with interferon-gamma in its cytotoxicity. This cytokine has been implicated in a variety of diseases, including autoimmune diseases, insulin resistance, and cancer.
Spécifications

Spécifications

P01375
4 pg/mL
ELISA
Human
Colorimetric Microplate Reader
CANDF6,IL-17 F,ML-1,ML1
HRP
Plasma 100 μL, Serum 100 μL, Supernatant 100 μL
Tumor Necrosis Factor alpha, tnf-a, tnf-alpha
1 hrs 45 mins
4 to 500 pg/mL
HRP
Plasma, Serum, Supernatant
ELISA
7124
Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, Biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, ELISA/ELISPOT Coating Buffer Powder: This uncoated ELISA kit may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 μM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20)., Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Streptavidin-HRP Concentrate (100X), Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86)
RUO
2°C to 8°C
TNF alpha
24 hrs 30 mins
Videos
FDS
Documents

Documents