Learn More
Invitrogen™ eBioscience™ TMB Solution (1X)
Catalog No. 501129758
Change view
Click to view available options
Quantity:
100 mL
Catalog No. Quantity
501129758 100 mL
1 options
Catalog No. 501129758 Supplier Invitrogen™ Supplier No. 00420156
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recommended for ELISA Ready-SET-Go!™ kits. eBioscience™ TMB ELISA Substrate Solution, 1x is a sensitive chromogenic substrate supplied as single ready-to-use solution.
TMB (3,3’,5,5’-Tetramethylbenzidine) is a substrate for horseradish peroxidase (HRP). During the enzymatic reaction the TMB substrate will result in a blue color that can be read on a spectrophotometer at 650nm. This kinetic reaction can be stopped using 1M phosphoric acid resulting in a color change to yellow that can be read at 450nm.
Specifications
Product Type TMB Solution
Quantity 100 mL
Detection Method Colorimetric
Product Line eBioscience
Substrate Properties Chemical Substrate
Substrate Type HRP (Horseradish Peroxidase)
For Research Use Only. Not for use in diagnostic procedures.