missing translation for 'onlineSavingsMsg'
Learn More

Titanium aluminide, 99.5% (metals basis)

Numéro de catalogue. AA2289514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA2289514 25 g
AA2289522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA2289514 Fournisseur Thermo Scientific Chemicals Code fournisseur 02289514
Il en reste null

Used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products. | CAS: 12004-78-3 | Al3Ti | 128.812 g/mol

Titanium aluminide is used in aircraft turbine engines, exhaust valves and turbochargers for passenger vehicle engines, intermediate process for some mill products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 12004-78-3
Molecular Formula Al3Ti
Molecular Weight (g/mol) 128.812
MDL Number MFCD00054094
InChI Key OQPDWFJSZHWILH-UHFFFAOYSA-N
Synonym titanium aluminum, aluminum, compd. with titanium 3:1, aluminum; titanium, aluminum, compound with titanium 3:1
PubChem CID 6336852
IUPAC Name aluminum;titanium
SMILES [Al].[Al].[Al].[Ti]

Spécifications

Physical Form Powder
Quantity 25 g
UN Number UN3178
Solubility Information Insoluble in water.
Percent Purity 99.5%
Packaging Plastic bottle
Chemical Name or Material Titanium aluminide
Hazard Category H228-H315-H319-H335
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: II
EINECSNumber 234-461-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only