Learn More
Titanium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Ti), LabChem™
Supplier: LabChem LC261501
Specifications
| Titanium AA Standard | |
| Poly Bottle | |
| 99.6,0.4 | |
| Liquid | |
| (NH4)2TiF6 | |
| 100°C | |
| 500 mL | |
| Certified |
| 1000ppm (1mL = 1mg Ti) | |
| 16962-40-6 , 7732-18-5 | |
| Passes Test | |
| 1000 ppm ±10 ppm (1mL = 1mg Ti) | |
| Colorless | |
| Soluble in water | |
| 197.93 |
Chemical Identifiers
| 16962-40-6 |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage : Room Temperature