accessibility menu, dialog, popup

UserName

NH4NH4FFFFFFTi

CAS RN 16962-40-6

IUPAC Name: titanium(4+) diammonium hexafluoride
Synonyms: titanium(4+) diammonium hexafluoride
Molecular Weight (g/mol): 197.94
Molecular Formula: F6H8N2Ti
InChi Key: NMGYKLMMQCTUGI-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[F-].[F-].[F-].[F-].[F-].[F-].[Ti+4]
Grade 
  • (6)
  • (2)
  • (9)
  • (1)
  • (3)
Solvent or Matrix 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Technique 
  • (7)
  • (7)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (14)
  • (9)
  • (6)
  • (6)
  • (2)
  • (9)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (7)
  • (7)
  • (2)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)