missing translation for 'onlineSavingsMsg'
Learn More

Titanium AA Standard, Certified, 1000ppm ±10ppm (1mL = 1mg Ti), LabChem™

Catalog No. p-5388688
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC261507 125 mL Poly Bottle
LC261501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC261507 Supplier LabChem Supplier No. LC261507
Only null left

Chemical Identifiers

CAS 16962-40-6 , 7732-18-5

Specifications

Packaging Poly Bottle
CAS Max % 99.6, 0.4
Identification Passes Test
Physical Form Liquid
Concentration 1000 ppm ±10 ppm (1mL = 1mg Ti)
Linear Formula (NH4)2TiF6
Color Colorless
Boiling Point 100°C
Solubility Soluble in water
Quantity 125 mL
Formula Weight 197.93
Grade Certified
Chemical Name or Material Titanium AA Standard, 1000ppm (1mL = 1mg Ti)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature