Learn More
Epredia™ TissueLoc™ Cassette IV Biopsy Cassettes
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Spécifications
Spécifications
| Pore Style | Small Squares |
| Quantity | 1000/Cs. |
| Closure Included | Yes |
| Closure Type | Separate Lid, Back Hinge |
| PrintMate Compatible | No |
| Color | Blue |
| Product Type | TissueLoc Cassette IV with Color-Matched Lids |