Learn More
Corning™ PYREX™ Three-Way Angle Connecting Adapter, Standard Taper Joint
Three-way connecting tube with side arm placed at 75° angle from lower joint
Supplier:  Corning™ 900014
 Afficher plus de versions de ce produit
Numéro de catalogue. 153234A
Il en reste null
Avertissement:
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Description
Description
The sidearm is at an angle approximately 75° from the lower joint. The lower end and sidearm are equipped with inner joints, and the upper end with an outer joint. All joints on each tube are of the same size.
Spécifications
Identifiants chimiques
Borosilicate Glass (3.3)
75°
Tube Adapter
Spécifications
Borosilicate Glass (3.3)
10.5 cm
75°
4.13 in.
Two 14/20 Inner Bottom/Outer Top
Tube Adapter
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS