Learn More
Corning™ PYREX™ Three-Way Angle Connecting Adapter, Standard Taper Joint
GREENER_CHOICE
Three-way connecting tube with side arm placed at 75° angle from lower joint
$253.53 - $257.90
Specifications
Material Borosilicate Glass (3.3)
Arm Angle 75°
Product Type Tube Adapter
View More Specs
Alert:
Because of variations in conditions, glassware cannot be guaranteed against breakage under vacuum or pressure. Adequate precautions should be taken to protect personnel who are doing such work.
Products 3
Catalog Number Mfr. No. Overall Length (English) Cone Size Price Quantity & Availability  
Catalog Number Mfr. No. Overall Length (English) Cone Size Price Quantity & Availability  
153234A
View Documents Promotion Details
Corning™
900014
4.13 in. Two 14/20 Inner Bottom/Outer Top
Case for $257.90
Only null left
153234
GREENER_CHOICE
View Documents Promotion Details
Corning™
900024
6.29 in. Two 24/40 Inner Bottom/Outer Top
Each for $253.53
Case of 6 Each for $1,521.18
Only null left
153234B
GREENER_CHOICE
View Documents Promotion Details
Corning™
900019
4.13 in. Two 19/22 Inner Bottom/Outer Top
Case for $256.98
Only null left
Description
Description
The sidearm is at an angle approximately 75° from the lower joint. The lower end and sidearm are equipped with inner joints, and the upper end with an outer joint. All joints on each tube are of the same size.
Specifications
Specifications
Borosilicate Glass (3.3)
75°
Tube Adapter