Learn More
Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for GC Instruments
Choose Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes for accurate, reproducible GC sample introduction using an affixed needle and a plunger that creates a tight seal.
$166.13 - $1,145.80
Specifications
Connection Type Fixed
View More Specs
Products 18
Catalog Number Mfr. No. Volume (Metric) Needle Gauge Length (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Needle Gauge Length (Metric) Quantity Price Quantity & Availability  
365D2976
View Documents
Thermo Scientific™
365D2976
10 μL 26 G 50 mm 6/Pk.
Pack of 6 Each for $1,145.80
365D2977
View Documents
Thermo Scientific™
365D2977
10 μL 26 G 50 mm 1/Pk.
Each for $166.13
365F3761
View Documents
Thermo Scientific™
365F3761
25 μL 26 G 50 mm 1/Pk.
Each for $293.02
365F3700
View Documents
Thermo Scientific™
365F3700
25 μL 26 G 50 mm 1/Pk.
Each for $529.69
365H3771
View Documents
Thermo Scientific™
365H3771
100 μL 23 G 50 mm 1/Pk.
Each for $302.25
365H5700
View Documents
Thermo Scientific™
365H5700
100 μL 26 G 50 mm 1/Pk.
Each for $344.95
365H6700
View Documents
Thermo Scientific™
365H6700
250 μL 26 G 50 mm 1/Pk.
Each for $452.77
365J7700
View Documents
Thermo Scientific™
365J7700
500 μL 26 G 50 mm 1/Pk.
Each for $433.77
365K8135
View Documents
Thermo Scientific™
365K8135
1 mL 26 G 50 mm 1/Pk.
Each for $648.39
365L8635
View Documents
Thermo Scientific™
365L8635
2.5 mL 26 G 50 mm 1/Pk.
Each for $703.73
365H2151
View Documents
Thermo Scientific™
365H2151
100 μL 26s G 85 mm 1/Pk.
Each for $437.70
03100046
View Documents
Thermo Scientific™
365D0621
10 μL 23 to 26 G 42 mm 1/Pk.
Each for $190.60
365H2171
View Documents
Thermo Scientific™
365H2171
100 μL 23 G 85 mm 1/Pk.
Each for $655.08
365J2411
View Documents
Thermo Scientific™
365J2411
500 μL 26 G 85 mm 1/Pk.
Each for $809.40
365J2421
View Documents
Thermo Scientific™
365J2421
500 μL 26 G 57 mm 1/Pk.
Each for $677.51
365J2441
View Documents
Thermo Scientific™
365J2441
500 μL 23 G 57 mm 1/Pk.
Each for $829.04
365D0631
View Documents
Thermo Scientific™
365D0631
Each for $195.34
365D0636
View Documents
Thermo Scientific™
365D0636
Pack for $1,078.02
Description
Description
Get durability, clarity and accuracy in gas chromatography (GC) sample introduction for confidence in analytical results, time-after-time. Thermo Scientific™ Fixed-Needle, Gas-Tight Syringes offer an affixed needle and a plunger that creates a tight seal with the barrel. Choose from a variety of sizes, types and gas chromatography autosampler compatibilities. They are available in standard industry size with black easy-to-read and solvent-resistant markings.
Durability
The enhanced design of the syringes delivers:
  • Improved solvent resistance
  • Greater temperature range
  • Increased operational smoothness
  • Longer lifetime and improved cycle life
Clarity
Syringes are provided with a clear, easy-to-read black scale, which aids in consistent delivery of samples for manual syringes and easy identification of installed autosampler syringes.
Accuracy
The enhanced design of the syringe has eliminated areas where the sample can become trapped and potentially cause carryover:
  • Closer fit between the plunger tip and PTFE insert at the zero position
  • Tighter fit between PTFE insert and glass barrel
  • Improved fixed needle attachment
In addition, the reduction in adhesive from the fluid path enhances durability and reduces the risk of sample interaction with the adhesive.
Specifications
Specifications
Fixed
Videos
SDS
Documents
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.