missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ WELQut Protease (5 U/μL)

Catalog No. FEREO0861
Change view
Click to view available options
Quantity:
500 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREO0861 500 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREO0861 Supplier Thermo Scientific™ Supplier No. EO0861

missing translation for 'validMainframeMsg'

Thermo Scientific™ WELQut Protease is a highly specific serine protease cleaving outside the WELQX recognition motif to remove N-terminal protein fusion tags.

Thermo Scientific WELQut Protease is highly specific, recombinant serine protease of Staphylococcus aureus. It recognizes and precisely cleaves recombinant proteins containing an engineered recognition sequence† W- E- L- Q↓X (Trp, Glu, Leu, Gln, X can be any amino acid). The protease cleaves outside the recognition sequence without leaving additional amino acids bound to the target protein.
The WELQut Protease is active in a broad temperature (4 to 30°C) and pH (pH 6.5 to 9.0) range and does not require specific buffers.
In addition, this new protease has several procedural advantages - it is ideal for on-column proteolysis reactions and can be easily removed from reaction mixtures using its built-in His-tag.

Highlights

  • Cleaves outside WELQ recognition sequence, without leaving additional amino acids bound to the target protein
  • Highly specific to cognate recognition site, does not generate non-specific product bands, even after long incubation and using excess of protease
  • Easy to remove from the reaction mixture using built-in His-tag
  • Ideal for on-column proteolysis reactions
Applications
  • Removal of N-terminal fusion tags from recombinant protein preparations.
Footnote
† This cleavage sequence is present in expression vector pLATE52 included into the Thermo Scientific aLICator LIC Cloning and Expression Kit 4 (N-terminal His-tag/WELQut) (#K1281) available from Thermo Scientific.

Definition of Activity Unit: Each unit is defined as the amount of enzyme required to cleave ≥ 99% of 100 μg of a control protein in 16 h at 20°C.

  • Enzyme activity is assayed in 100 μL 100 mM Tris-HCl (pH 8.0)
  • Hazardous: No
  • Molecular Weight: 22 kDa monomer
  • Source:Bacillus subtilis cells with a cloned gene of SplB protease from Staphylococcus aureus
  • Storage Buffer: Enzyme is supplied in 10 mM Na2HPO4; 1.8 mM KH2PO4, pH 7.3; 140 mM NaCl; 2.7 mM KCl, 50% glycerol
  • Storage Condition:-20°C
  • -20°C

Specifications

Product Line WELQut
Quantity 500 units
Product Type Protease

For Research Use Only. Not for use in diagnostic procedures.