missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium ICP Standard, 10,000 ppm Tl in 5% HNO3, Ricca Chemical
$1122.13 - $1122.13
Chemical Identifiers
| CAS | 7440-28-0 |
|---|---|
| Molecular Formula | Tl |
| Molecular Weight (g/mol) | 204.38 |
| MDL Number | MFCD00134063 |
| InChI Key | BKVIYDNLLOSFOA-UHFFFAOYSA-N |
| PubChem CID | 5359464 |
| ChEBI | CHEBI:30440 |
| IUPAC Name | thallium |
| SMILES | [Tl] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PTL10KN-500
|
Ricca Chemical Company
PTL10KN500 |
500 mL | Natural Poly Bottle |
Each for $1,122.13
|
|
||||
Chemical Identifiers
| 7440-28-0 | |
| 204.38 | |
| BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
| CHEBI:30440 | |
| [Tl] |
| Tl | |
| MFCD00134063 | |
| 5359464 | |
| thallium |
Specifications
| Natural Poly Bottle | |
| 10,000ppm Tl | |
| 89.0,12.05,0.89 | |
| Liquid | |
| MFCD00134063 | |
| < 2 | |
| BKVIYDNLLOSFOA-UHFFFAOYSA-N | |
| thallium | |
| 500 mL | |
| CHEBI:30440 |