missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep Group LLC PLTB22X
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Tb | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
| Terbium Standard | |
| 7440-27-9 , 7697-37-2 , 7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 1.0,5.0,94.0 | |
| MFCD00011256 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| GZCRRIHWUXGPOV-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 158.93 | |
| 23958 | |
| Assurance |
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| 1.0,5.0,94.0 | |
| Tb | |
| AA, ICP | |
| Certified Reference Material | |
| [Tb] | |
| terbium | |
| 500 mL | |
| CHEBI:33376 | |
| Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLTB2-2Y, -2X 10,000 PLTB2-3Y, -3X |
Chemical Identifiers
| 7440-27-9 | |
| 158.93 | |
| GZCRRIHWUXGPOV-UHFFFAOYSA-N | |
| CHEBI:33376 | |
| [Tb] |
| Tb | |
| MFCD00011256 | |
| 23958 | |
| terbium |
Safety and Handling
Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only