CAS RN 7440-27-9
Terbium pieces, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium ingot, 99.9% (REO), Ta <0.4%
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium powder, -40 mesh, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium sputtering target, 50.8mm dia x 3.18mm thick, Thermo Scientific™
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium foil, 0.62mm (0.024in) thick, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium rod, 12.7mm (0.50in) dia, 99.9% (metals basis excluding Ta)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium pieces, distilled dendritic, 99.9% (REO)
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N Synonym: unii-06ssf7p179,ingot,foil,terbio,atom,chips,pieces,powder,acmc-20ajly,pieces, distilled dendritic PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]
Terbium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Terbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-27-9 Molecular Formula: Tb Molecular Weight (g/mol): 158.93 MDL Number: MFCD00011256 InChI Key: GZCRRIHWUXGPOV-UHFFFAOYSA-N PubChem CID: 23958 ChEBI: CHEBI:33376 IUPAC Name: terbium SMILES: [Tb]